Learn More
Gibco™ IMDM

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells
£19.31 - £116.00
Specifications
Cell Line | Jurkat, COS-7, and macrophage cells |
---|---|
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Product Type | IMDM (Iscove's Modified Dulbecco's Medium) |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11504556
![]() |
Gibco™
12440053 |
500 mL |
£19.31
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12589059
![]() |
Gibco™
12440061 |
10 x 500 mL |
£116.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10135083
![]() |
Gibco™
12440046 |
1000 mL |
£23.11
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This IMDM is modified as follows:
With: L-glutamine, Phenol Red
Without: α-thioglycerol, 2-mercaptoethanol
The complete formulation is available.
Using IMDM
IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Jurkat, COS-7, and macrophage cells | |
1 X | |
IMDM (Iscove's Modified Dulbecco's Medium) | |
Sterile-filtered | |
No α-Thioglycerol, No 2-Mercaptoethanol | |
Gibco™ | |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For in vitro diagnostic use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.