missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Kanamycin Sulfate
Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus
Brand: Gibco™ 11815024
35.30 GBP valid until 2024-12-31
Use promo code "21971" to get your promotional price.
This item is not returnable.
View return policy
Description
- A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
- Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
- Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
- It is used for the prevention of bacterial contamination of cell cultures
- Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
- Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
- Store at -5°C to -20°C
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Store at room temperature. | |
Antibiotic | |
5 g |
Powder | |
Bacterial Selection | |
Ambient Temperature |
For Research Use Only. Not for use in diagnostic procedures.