Learn More
Gibco™ LV-MAX™ Production Medium
£205.00 - £1560.00
Specifications
Content And Storage | Store at 2–8°C, protect from light. |
---|---|
Cell Line | Viral Production Cell-Line |
Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
Culture Type | Suspension Cell Culture |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15863374
|
Gibco™
A3583401 |
1 L | N/A | ||||||
15839081
|
Gibco™
A3583401 |
1 L |
£285.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15849081
|
Gibco™
A3583402 |
6 x 1L |
£1560.00
6 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco LV-MAX Production Medium is a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells (suspension-adapted HEK 293-derived cells) promoting high-titer lentiviral production.
- Supports growth of suspension Viral Production Cells to densities of over 1.0 x 107 cells/mL
- Enables sustained, high-level production of high-density transiently transfected cultures, achieving titer of up to 1 x 108 TU/mL (unconcentrated LVVs-GFP)
- Supports growth and transfection of Viral Production Cells in culture formats from 96-deep well blocks to 2-L flasks and bioreactors
LV-MAC Production Medium is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium is not recommended for adherent cell cultures.
Specifications
Store at 2–8°C, protect from light. | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Liquid | |
No antibiotics | |
Room Temperature |
Viral Production Cell-Line | |
Suspension Cell Culture | |
Lentiviral Production Medium | |
LV-MAX™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.