Learn More
Gibco™ McCoy's 5A (Modified) Medium
McCoy's 5A (modified) Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.
£35.31
Specifications
Content And Storage | Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | Rat fibroblasts |
Cell Type | Biopsy Tissues |
Classification | Animal Origin |
Concentration | 1 X |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410604
|
Gibco™
16600082 |
500 mL |
£35.31
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
McCoy's 5A (modified) Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.
This McCoy's 5A is modified as follows:
With: High Glucose, L-glutamine, Bacto-peptone, Phenol Red
Without: Sodium Pyruvate, HEPES
The complete formulation is available.
Using McCoy's 5A (modified) Medium
Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator. McCoy's 5A (modified) Medium requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). McCoy's 5A (modified) Medium uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Biopsy Tissues | |
1 X | |
McCoy's 5A Modified Medium | |
No HEPES, No Sodium Pyruvate | |
Gibco™ |
Rat fibroblasts | |
Animal Origin | |
Liquid | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
Your input is important to us. Please complete this form to provide feedback related to the content on this product.