missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM
GREENER_CHOICE

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media.

Brand:  Gibco™ 11095080

17.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15188319

  • £19.95 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

MEM (Minimum Essential Medium) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With: L-glutamine, Phenol Red

Without: HEPES

The complete formulation is available.

Using MEM

MEM was developed by Harry Eagle, based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts. MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No HEPES, No Sodium Pyruvate
Gibco™
Room Temperature
Primary Rat Astrocytes
1 X
MEM (Minimum Essential Medium)
Sterile-filtered
Low Glucose, Glutamine, Phenol Red
cGMP-compliant under the ISO 13485 standard
500 mL
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2537078 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11095080
Lot Number2646171 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11095080
Lot Number2537070 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11095080
Lot Number2645336 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11095080
Lot Number2522611 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11095080
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ MEM > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.