missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf-900™ II SFM
GREENER_CHOICE

Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells

Brand:  Gibco™ 10902104

427.00 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10003502

  • £610.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Sf-900 II SFM is a serum-free, protein-free insect cell culture medium for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.

  • Superior long-term, high-density growth
  • Optimized for recombinant protein production
  • Serum-free, protein-free, ready-to-use formulation
  • Scalable in bioreactors

Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Insect Cell
Liquid
Serum-free
Sustainable packaging
6 x 1 L
Sf21, Sf9
Protein-free, Serum-free
Insect Cell Serum Free Medium (SFM)
Glutamine
Gibco™, Sf-900™
S. frugiperda, Spodoptera frugiperda
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.