Learn More
Gibco™ Sf-900™ II SFM
Optimized for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS)
Brand: Gibco™ 10902179
Description
Sf-900 II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells. This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.
- Superior long-term, high-density growth
- Optimized for recombinant protein production
- Serum-free, protein-free, ready-to-use formulation
- Scalable in bioreactors
Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Insect Cell | |
Liquid | |
Serum-free | |
Gibco™, Sf-900™, SynerGy™ | |
Room Temperature |
Sf21, Sf9 | |
Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
Glutamine | |
10 L | |
S. frugiperda, Spodoptera frugiperda |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.