Learn More
Gibco™ Sf-900™ II SFM
Optimized for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS)
£410.00 - £1950.00
Especificaciones
Content And Storage | Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | Sf21, Sf9 |
Cell Type | Insect Cell |
Classification | Protein-free, Serum-free |
Form | Liquid |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
10679234
|
Gibco™
10902179 |
10 L |
£1070.00
Each |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11390872
|
Gibco™
10902187 |
20 L |
£1950.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10680745
|
Gibco™
10902161 |
5 L |
£410.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Sf-900 II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells. This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.
- Superior long-term, high-density growth
- Optimized for recombinant protein production
- Serum-free, protein-free, ready-to-use formulation
- Scalable in bioreactors
Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Información de órdenes
Shipping Condition: Room temperature
Especificaciones
Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Insect Cell | |
Liquid | |
Serum-free | |
Gibco™, Sf-900™, SynerGy™ | |
S. frugiperda, Spodoptera frugiperda |
Sf21, Sf9 | |
Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
Glutamine | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.