Learn More
Gibco™ TrypLE™ Express Enzyme (1X), no phenol red

Unlike porcine trypsin, free of animal-derived components
£24.05 - £684.00
Specifications
Chelators | EDTA |
---|---|
Cell Type | Mammalian Cells |
Classification | Animal Origin-free |
Concentration | 1 X |
Culture Type | Adherent Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10718463
![]() |
Gibco™
12604013 |
100 mL |
£24.05
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11528856
![]() |
Gibco™
12604021 |
500 mL |
£79.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11538856
![]() |
Gibco™
12604039 |
20 x 100mL |
£360.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11570926
![]() |
Gibco™
12604054 |
5 L |
£684.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.
- Gentle on cells
- Room temperature-stable for 24 months
- Easy to use
- Animal origin-free
- Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
- Can be directly substituted for trypsin in existing protocols
- Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP-compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
EDTA | |
Animal Origin-free | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
No Phenol Red | |
Check and Record | |
Room Temperature Stable | |
Animal origin-free |
Mammalian Cells | |
1 X | |
Liquid | |
Sterile-filtered | |
Energy efficient, Sustainable packaging | |
Gibco™ | |
Room Temperature | |
In vitro Bioassay |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.