Learn More
Gibertini Elettronica™ Digital Tensiometer
Measure liquid surface tension with efficiency, using this tensiometer. Gibertini Elettronica™ Digital Tensiometers feature a simple, robust design for fast response times and accurate results.
Brand:  Gibertini Elettronica™ TSD
Encompass
Product Code. 11900877
Description
Description
  • Glass plates (dimensions 24 x 24 x 0.15mm)
  • Glass container for liquid sample
  • Manual adjustable lab jack
  • Suspended system for the arrangement of glass plate
  • Available with MISTEL automated jack, and WIND DCA 300 software to pilot the jack and automatically determine the surface tension and contact angle requirement
Specifications
Specifications
Digital Tensiometer
Measuring liquid surface tension
Glass plate dimensions: 24 x 24 x 0.15mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Gibertini Elettronica™ Digital Tensiometer