Learn More
Ambion™ Customer-Defined Pre-miR™ miRNA Precursor
Customer-Defined Pre-miR™ miRNA Precursor
Brand: Ambion™ AM17103
597.64 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. Use our customization tool to design the ideal mimic for your miRNA sequence, or select from among predesigned products.
Specifications
Room temperature | |
Pre-miRNA Precursor, 20nmol, Small, Chemically Modified Double-Stranded RNA Molecules Designed to Mimic Endogenous Mature miRNAs, Use Our Customization Tool to Design the Ideal Mimic for Your miRNA Sequence | |
Pre-miRNA Precursor | |
20 nmol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Ambion™ Customer-Defined Pre-miR™ miRNA Precursor > 20 nmol