Learn More
Gilson™ PIPETMAN™ Classic Pipet
GREENER_CHOICE
The PIPETMAN Classic line, the world standard in pipetting, offers a wide range of continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density.
Brand:  Gilson™ F123615
 View more versions of this product
Product Code. 10442412
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability
Order Info
Trade in program available. Please contact your Fisher Scientific Sales representative for more details.
Specifications
Specifications
±0.35, ±1.8μL
No
Pink
Reusable
Polypropylene
1
No
Pipette
Small volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology)
36.5 cm
30.5 cm
12 cm
Certified Quality
D200, DF100ST
Autoclavable (Tip Holder and Tip Ejector)
Manual
Ergonomic
P100
Pipetman™ Classic™
Non-sterile
20 to 100 μL
14.37 in.
12.01 in.
4.72 in.
Pink
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products