Learn More
Gilson™ PIPETMAN™ EcoPack™ Tips
GREENER_CHOICE
Ensures optimal performance of PIPETMAN pipettes across the full volume range. Gilson™ PIPETMAN™ EcoPack™ Tips are fully autoclavable.
Brand:  Gilson™ F161930
167.51 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 12507536
  • £289.00 / Pack of 10000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk Packs are sealed with a tamper-proof strip which can easily be opened for quick access.
  • Bulk Packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EcoPack box contains 10,000 tips (10 bags of 1000 tips).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 200 μL
Non-sterile
Autoclavable
23.62 in.
Bagged
Pipet Tips
Single-use
Yes
Polypropylene
Non-filtered
Clear
D200
60 cm
Universal
Yellow
Gilson Pipetman
Pipetman EcoPack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ PIPETMAN™ EcoPack™ Tips > PIPETMAN™ EcoPack™ Tips; Volume: 2 to 200 μL; Quantity: 1000 Tips/Box