missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ QC System v2

The RNaseAlert™ QC System v2 detects RNase activity in convenient and sensitive fluorimetric assay that delivers results in real time.

Brand:  Invitrogen™ 4479769


Product Code. 15286535

  • £1036.00 / 480 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The RNaseAlert™ QC System v2 detects RNase activity in convenient and sensitive fluorimetric assay that delivers results in real time. This version contains substrate that is ∼60% more sensitive than previous version. The kit contains sufficient reagents for 5 x 96 (480) high-throughput assays.

  • Detects as little as approx. 0.3 pg of RNase A with fluorometer
  • Simple, straight-forward, 30-minute assay
  • Designed to monitor RNase activity in column fractions during protein purificatio
  • Use with DNaseAlert™ QC System to quantitatively detect both RNases and DNases in same sample
  • Compatible with DNaseAlert QC System
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C:
• 5 tubes of RNaseAlert™ Substrate v2
• 5 mL 10X RNaseAlert Lab Test Buffer
• 500 μL RNase A

Store at room temperature:
• 250 mL RNaseZap™ Solution

Store at any temperature:
• 50 mL Nuclease-free Water

RNase Control Reagent
RNA Extraction
480 reactions
RNaseAlert™
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2847698 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number4479769
Lot Number2816340 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479769
Lot Number2823216 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479769
Lot Number2815563 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479769
Lot Number2805190 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4479769
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ RNaseAlert™ QC System v2 > 480 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.