missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNaseAlert™ QC System

Provides sensitive detection of DNases in a simple, easy-to-use fluorimetric assay with real time results

Brand:  Invitrogen™ AM1970

Product Code. 10594825

  • £866.00 / Pack of 480

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyophilized
Dry Ice
DNase Control Reagent

Store DNaseAlert™,10X NucleaseAlert Buffer, DNase I and TE Buffer pH 7.0 at -20°C.

Store RNaseZap™Solution, Non-stick Tubes and Nuclease-free Water at room temperature.

480 assays
Ambion™, RNaseAlert™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2790583 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberAM1970
Lot Number2828008 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1970
Lot Number2798257 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1970
Lot Number2827730 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1970
Lot Number2804727 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1970
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ DNaseAlert™ QC System > 480 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.