Learn More
Gilson™ PIPETMAN™ L Multichannel Pipet
GREENER_CHOICE
The PIPETMAN™ L Multichannel Pipet is a lightweight multichannel pipet with lockable volume that has been designed to fit naturally in your hand.
Brand:  Gilson™ FA10016
991.26 GBP valid until 2025-12-31
Use promo code "24315" to get your promotional price.
 View more versions of this product
Product Code. 13446718
  • £1120.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • Redesigned handle fits naturally in your hand, no matter the size.
  • Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
  • 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipet.
  • Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No
Green
Reusable
Polypropylene
12
No
Pipette
Ergonomic
6.02 in.
12.87 in.
1.5 in.
0.92 lb.
Certified Quality
Autoclavable
Manual
Ergonomic
P12x300L
Pipetman L
Non-sterile
20 to 300 μL
1 Year
15.3 cm
32.7 cm
3.8 cm
Green
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products