Learn More
Gilson™ PIPETMAN™ TIPACK™ Racked Pipet Tips
GREENER_CHOICE
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ TIPACK™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Brand:  Gilson™ F171300
49.57 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 12649581
  • £69.45 / Pack of 960
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Requires low tip-mounting force.
  • Robust and designed to withstand autoclaving.
  • Sterility assurance level SAL 10-6 guaranteed, using gamma irradiation.
  • All packs are wrapped in medical device quality plastic which provides a complete shield against contamination.
  • Indicator dots used on Gilson products undergo an irreversible color change upon sterilization; yellow gamma dots turn red when products have been properly exposed.
  • TIPACKs are securely sealed with a sticker; a full product description is given on the TIPACK for traceability and easy identification.
  • Available in sterilized or autoclavable packages.
  • Manufactured in a clean room and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging to meet GLP requirements.
Order Info
10 TIPACKs of 96 racked tips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 200 μL
Non-sterile
Autoclavable
5.71 in.
Rack
Single-use
Polypropylene
Non-filtered
Clear
D200
14.5 cm
Pipet Tips
Pipetman Tipack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Master liquid handling with ease.
    Promo Code: 24122
    Expires2025-04-13
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Gilson™ PIPETMAN™ TIPACK™ Racked Pipet Tips > PIPETMAN™ TIPACK™ Racked Pipet Tips; Model: D200; Autoclavable; Volume: 2 to 200 μL; Length (Tip): 51 mm