missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GeneBLAzer™ GeneBLAzer™ GIPR-CRE-bla HEK 293T Cells
GeneBLAzer™ GIPR-CRE-bla HEK 293T Cells contain the human Gastric Inhibitory Peptide Receptor (GIPR).
Brand: GeneBLAzer™ K1864
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Accession Number: NM_000164.2
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ GIPR-CRE-bla HEK 293T cells contain the human Gastric Inhibitory Peptide Receptor (GIPR), stably integrated into the CellSensor™ CRE-bla HEK 293T cell line. CellSensor™ CRE-bla HEK 293T cells (Cat. no. K1535)) contain a beta-lactamase reporter gene under control of the CRE Response Element. The GeneBLAzer™ GIPR-CRE-bla HEK 293T cells are functionally validated for Z’-factor and EC50 concentrations of Gastric Inhibitory Peptide (GIP). In addition, GeneBLAzer™ GIPR-CRE-bla HEK 293T cells have been tested for assay performance under variable conditions.
Specifications
NM_000164.2 | |
Dividing Cells | |
Fluorescent | |
G-Protein Coupled Receptor (GPCR) | |
GeneBLAzer™ | |
GIPR | |
293T (HEK) | |
GPCR |
Cell-based beta lactamase reporter gene | |
GeneBLAzer™ GIPR-CRE-bla HEK 293T cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
FRET | |
Single Cell Clone Origin | |
End Point | |
Cell Line | |
GeneBLAzer™ |
For Research Use Only. Not for use in diagnostic procedures.