missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bulb Pipettes, One Mark, Class B, BS 1583

Pipette with precision, Fisherbrand one mark Class B pipettes are made of soda lime glass. Colour coded with blue markings.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.21060kg

 View more versions of this product

Product Code. 11962168

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Color coded to BS 3996
  • Class B
  • Soda Lime Glass
  • Graduated
  • One Mark
  • Type 1
Specifications

Specifications

To Deliver
B
Yes
±0.06 mL
10 mL
ISO 648, DIN 12690, Class B
Soda Lime Glass
5 Pack
Red
Bulb Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers