missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Pear-Shaped Separating Funnel with PTFE Stopcock

Constructed in chemical resistant borosilicate glass. Fisherbrand™ Glass Pear-Shaped Separating Funnel with PTFE Stopcock separates mixtures into immiscible solvents. Available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.

Brand:  Fisherbrand™ FB51759

141.92 GBP valid until 2024-12-31
Use promo code "21757" to get your promotional price.


 View more versions of this product

Product Code. 11982138

  • £227.00 / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Constructed in chemical resistant borosilicate glass. Fisherbrand™ Glass Pear-Shaped Separating Funnel with PTFE Stopcock separates mixtures into immiscible solvents. Available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.

  • Constructed in chemical resistant borosilicate glass.
  • ISO 4800 Certified.
    • Easy-open stopper helps to reduce pressure buildup during processing.
    • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    1000 mL
    ISO 4800
    PTFE
    29/32
    Borosilicate Glass
    1PK
    Glass, Plastic
    Separatory Funnel
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.