Learn More
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
Brand:  Poulten Graf™ 101 08153
 View more versions of this product
Product Code. 10519111
  • £1470.00 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For dispensing of pure organic solvents. Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • Solid dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Fortuna Optifix Solvent Dispenser
40 to 200 mL
Each
PTFE, Polypropylene, Fiberglass
For dispensing solvents
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser > Volume: 40 to 200 mL