Learn More
Hach Lange™ Glass Sample Cells
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
Brand:  Hach Lange™ 2084900
 View more versions of this product
Product Code. 10132114
  • £92.05 / Pack of 6
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
  • For use with Hach Ratio 18900, 2100N and 2100AN Laboratory Turbidmeters
  • Replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters
  • For use with Hach Ratio 18900, 2100N and 2100AN laboratory turbidmeters
  • Includes six sample cell vials with caps
  • Made of glass material\
Specifications
Specifications
Glass Sample Cell
2100N, 2100AN, and Lab RATIO Turbidimeters
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hach Lange™ Glass Sample Cells > For use with: 2100N, 2100AN, and Lab RATIO turbidimeters