missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Glass Serological Pipet

Brand:  Poulten Graf™ D806

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12374438

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Glass
0.02 mL
2 mL
Serological Pipette
Yes
500/Pk.
260 mm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers