Learn More
Hirschmann™ Glass Trapezoidal Volumetric Flask Class A with Glass Stopper
For volumetric accuracy and precision in liquid handling.
£43.75 - £47.70
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
13147959
|
Hirschmann™
2960253 |
5 mL | NS 10/19 |
£43.75
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11734106
|
Hirschmann™
2960240 |
2 mL | NS 7/16 |
£45.43
Pack of 2 |
Estimated Shipment: 31-10-2024
Log in to see stock. |
||||
11734116
|
Hirschmann™
2960252 |
5 mL | NS 7/16 |
£45.45
Pack of 2 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||||
13157959
|
Hirschmann™
2960261 |
10 mL | NS 10/19 |
£44.09
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11798094
|
Hirschmann™
2960260 |
10 mL | NS 7/16 |
£45.45
Pack of 2 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||||
11724116
|
Hirschmann™
2960233 |
1 mL | NS 7/16 |
£45.43
Pack of 2 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
11708104
|
Hirschmann™
2960265 |
20 mL | NS 10/19 |
£47.70
Pack of 2 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
11718104
|
Hirschmann™
2960270 |
25 mL | NS 10/19 |
£47.70
Pack of 2 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
Description
- Class A
- Manufactured from DURAN™ borosilicate glass
- Blue graduation marks
- Certified conformity, with dated batch identification
- Trapezoidal shape
- Standard ground and glass stopper
- Calibrated for IN
- Acc. to DIN EN ISO 1042
Specifications
DIN EN ISO 1042 | |
A |