Learn More
Fistreem International™ Glass Water Still, Loughborough
Reliable and robust water still with a long service history. Fistreem International™ Glass Water Still, Loughborough is a traditional design that has served countless laboratories worldwide for many years.
Brand: Fistreem International™ WGS-560-040G
Code : 11
Additional Details : Weight : 5.00000kg
Description
This conventional style water still is available in two output sizes, 4L/hr and 8L/hr. Each produces a high quality pyrogen free distillate. The assembly and operation of these stills is simple and their well proven design ensures that the minimum of time is spent on their maintenance. The heavy duty chromium plated heaters are fitted with a thermostatic cut out which prevents damage to the glassware should the water supply be broken or fail completely. The cut out automatically resets when cool. A constant level device automatically maintains the boiler contents at the optimum level. A siphon tube fitted to the boiler enables easy descaling when necessary. With a bench/wall mounting stand.Specifications
240 V 50 Hz 3 kW | |
450 x 390 x 820 mm | |
4 L/hr. |
Water Still, Loughborough | |
240 V |