Learn More
Glassy carbon plate, 3mm (0.1 in.) thick, type 2, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
Brand: Thermo Scientific Alfa Aesar 042820.FI
Code : NEW
Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon plate | |
Tube | |
50 x 50 mm |
7440-44-0 | |
MFCD00133992 | |
Insoluble in water. |
RUO – Research Use Only