Learn More
Glassy carbon plate, 3mm (0.1 in.) thick, type 2, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
POA - £560.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11370008
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042820.FF
25 x 25 mm
£174.00
25mm
View alternative products
Save Up to 
»
In Stock
to see stock.
11346159
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042820.FI
50 x 50 mm
£560.00
50mm
View alternative products
Save Up to 
»
In Stock
to see stock.
11360008
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042820.GH
100 x 100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.