missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ 4B Media

Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.

Brand:  Cytiva 17-0756-01


 View more versions of this product

Product Code. 11594935

  • £268.00 / 10mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glutathione Sepharose 4 B
4 to 13
90 μm
10 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Glutathione Sepharose 4B, 10 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.