missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722
£61.60 - £2090.00
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
10579570
|
Fisher Chemical
G/0650/08 |
500 mL | HDPE plastic bottle |
£61.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12656707
|
Fisher Chemical
G/0650/15 |
1 L | HDPE plastic bottle |
£114.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10795711
|
Fisher Chemical
G/0650/17 |
2.5 L | HDPE plastic bottle |
£271.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12666707
|
Fisher Chemical
G/0650/21 |
5 L | HDPE plastic bottle |
£498.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214580
|
Fisher Chemical
G/0650/25 |
25 L | Plastic container |
£2090.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722
Identificatori chimici
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifica
56-81-5 | |
5 | |
C3H8O3 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
OCC(O)CO | |
92.09 | |
3.17 | |
92.09g/mol | |
Very Viscous Liquid |
18°C | |
290°C | |
MFCD00004722 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
0.003mbar 50 |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto