missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, puriss. p.a., ACS reagent, Honeywell
£27.01 - £209.93
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15685690
|
Honeywell Riedel-de Haën
49770-250ML |
250 mL |
£27.01
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15665690
|
Honeywell Riedel-de Haën
49770-1L |
1 L |
£104.76
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15675690
|
Honeywell Riedel-de Haën
49770-2.5L |
2.5 L |
£209.93
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
160°C (320°F) | |
C3H8O3 | |
MFCD00004722 | |
635685 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
1 mmHg (20°C) | |
ACS/puriss. p.a. |
1.25g/mL | |
698°F | |
HOCH2CH(OH)CH2OH | |
NONH for all modes of transport | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
OCC(O)CO | |
92.09 | |
3.1 (vs air) | |
92.09g/mol | |
≥99.5% (GC) | |
Glycerol |
Safety and Handling
missing translation for 'einecsNumber' : 200-289-5