missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, puriss. p.a., ACS reagent, Honeywell Product Code.: 15675690

Glycerol, puriss. p.a., ACS reagent, Honeywell

Product Code. 15675690
2.5 L
Click to view available options
Quantity:
250 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15675690

Brand: Honeywell Riedel-de Haën 497702.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.25g/mL
Flash Point 160°C (320°F)
Ignition Point 698°F
Linear Formula HOCH2CH(OH)CH2OH
Quantity 2.5 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Glycerol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, puriss. p.a., ACS reagent, Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.