missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Multi-Compendial, U.S.P., J.T. Baker™
£234.00 - £663.00
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15587844
|
Avantor J.T.Baker
0582-01 |
500 g | Wide Mouth Amber Glass Bottle |
£234.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15597844
|
Avantor J.T.Baker
0582-05 |
2 kg | Wide Mouth Amber Glass Bottle |
£663.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Specifications
56-40-6 | |
MFCD00008131 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
750 | |
75.07 | |
Multi-Compendial/USP |
C2H5NO2 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
NCC(O)=O | |
75.07 | |
CHEBI:15428 | |
1L = 1.16kg | |
Glycine |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glycine, Multi-Compendial, U.S.P., J.T. Baker™