Learn More
Thermo Scientific™ UltraLink™ Hydrazide Resin
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.
Highlights:
High-capacity – immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin
Fast and efficient – immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
Specific immobilization – the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate
Stable attachment – resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required
Preserves antibody function – immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
Convenient sizes – choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns
UltraLink Hydrazide Resin:
- Ligand: hydrazide
- Support: Thermo Scientific UltraLink Biosupport
- Spacer arm: 22 atoms
- Loading: activated with at least 15μmol hydrazide per milliliter of settled resin
- Format: 50% resin slurry in 0.02% sodium azide
Includes:
Kits contain GlycoLink Columns; coupling buffer, coupling catalyst and wash buffer; oxidizing reagent; desalting columns, PBS buffer pack; and column accessories
Recommended for:
Purify antigens using covalently coupled antibodies; Determine binding partners for a glycoprotein of interest; Perform large-scale affinity purifications
Specifications
Specifications
| Capacity (Metric) | 1 to 10 mg Oxidized Antibody or Glycoprotein Immobilizing per mL Resin |
| Description | UltraLink Hydrazide Resin |
| Content And Storage | Store at 4°C. |
| Product Line | UltraLink |
| Quantity | 10 mL |
| Type | Resin |
| Stationary Phase | Hydrazide |
| Format | Bottle |
| Column Type | Affinity, UltraLink™ Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.