missing translation for 'onlineSavingsMsg'
Learn More
Gold(I) chloride, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11329548

Gold(I) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11329548
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11329548

Brand: Thermo Scientific Alfa Aesar 043365.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold(I) chloride is used as a starting material for the preparation of Phosphine ligated gold(I) complexes, which is used as catalysts in synthetic chemistry. It is also used to prepare metallic gold and potassium auric bromide and potassium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special gradeShow MoreShow Less
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Specifications

Melting Point 289°C (decomposition)
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,4515
Solubility Information Soluble in hydrochloric acid,hydrobromic acid and organic solvents. Slightly soluble in water.
Formula Weight 232.42
Percent Purity 99.9%
Odor Odorless
Sensitivity Moisture sensitive
Density 7.57 g/mL
Assay (metals basis)
Chemical Name or Material Gold (I) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU06K067 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog Number043365.03,043365.06
Lot NumberP08I041 Certificate TypeCertificate of Analysis Date15/04/2022 Catalog Number043365.03,043365.06
Lot NumberT20H046 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043365.03,043365.06
Lot NumberP31D057 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043365.03,043365.06
Lot NumberI28L18 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043365.03,043365.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold(I) chloride, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.