missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) iodide, 99%, Thermo Scientific Chemicals

£188.94 - £811.04

Chemical Identifiers

CAS 10294-31-2
Molecular Formula AuI
Molecular Weight (g/mol) 323.87
MDL Number MFCD00014175
InChI Key ATGIETUGWDAYPU-UHFFFAOYSA-M
Synonym gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
PubChem CID 82526
IUPAC Name iodogold
SMILES [I-].[Au+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307198
View Documents
Thermo Scientific Alfa Aesar
016617.03
1 g
£188.94
1g
Please to purchase this item. Need a web account? Register with us today!
11397188
View Documents
Thermo Scientific Alfa Aesar
016617.06
5 g
£811.04
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold(I) iodide is employed in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10294-31-2
323.87
ATGIETUGWDAYPU-UHFFFAOYSA-M
82526
[I-].[Au+]
AuI
MFCD00014175
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
iodogold

Specifications

120°C (decomposition)
Crystalline
MFCD00014175
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
ATGIETUGWDAYPU-UHFFFAOYSA-M
iodogold
82526
99%
8.25 g/mL
10294-31-2
AuI
UN3260
Insoluble in water; Soluble in alkali iodide, cyanide solutions; Decomposes in warm acids
[I-].[Au+]
323.87
323.87
Moisture sensitive
Gold(I) iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 233-656-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only