Learn More
Gold(III) bromide, 99%, Thermo Scientific Chemicals
Description
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Specifications
Specifications
Melting Point | ∼160°C (decomposition) |
Quantity | 1 g |
UN Number | UN3260 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 436.69 |
Percent Purity | 99% |
Sensitivity | Hygroscopic |
Chemical Name or Material | Gold(III) bromide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253438 | Certificate of Analysis | 16/12/2024 | A19514.03,A19514.06 |
10240584 | Certificate of Analysis | 21/05/2022 | A19514.03,A19514.06 |
10238341 | Certificate of Analysis | 12/04/2022 | A19514.03,A19514.06 |
P16C016 | Certificate of Analysis | 23/11/2021 | A19514.03,A19514.06 |
F25S007 | Certificate of Analysis | 23/11/2021 | A19514.03,A19514.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.