missing translation for 'onlineSavingsMsg'
Learn More
Gold(III) bromide, 99%, Thermo Scientific Chemicals Product Code.: 11414087

Gold(III) bromide, 99%, Thermo Scientific Chemicals

Product Code. 11414087
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11414087

Brand: Thermo Scientific Alfa Aesar A19514.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-28-7
Molecular Formula AuBr3H3
Molecular Weight (g/mol) 439.70
MDL Number MFCD00014171
InChI Key KJCSJLXVPHOFJC-UHFFFAOYSA-N
IUPAC Name gold(III) bromide
SMILES Br.Br.Br.[Au]

Specifications

Melting Point ∼160°C (decomposition)
Quantity 5 g
UN Number UN3260
Solubility Information Slightly soluble in water.
Formula Weight 436.69
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material Gold(III) bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253438 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA19514.03,A19514.06
Lot Number10240584 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA19514.03,A19514.06
Lot Number10238341 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA19514.03,A19514.06
Lot NumberP16C016 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19514.03,A19514.06
Lot NumberF25S007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19514.03,A19514.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold(III) bromide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.