missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold wire, 0.25mm (0.01 in.) dia., 99.9% (metals basis), Thermo Scientific Chemicals
£237.00 - £1137.00
Specifications
Chemical Name or Material | Gold wire |
---|---|
CAS | 7440-57-5 |
Assay Percent Range | 99.9% (metals basis) |
Assay Percent Notes | (metals basis) |
Molecular Formula | Au |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396627
|
Thermo Scientific Alfa Aesar
000725.G1 |
1 m |
£237.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386627
|
Thermo Scientific Alfa Aesar
000725.G5 |
5 m |
£1137.00
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold wire | |
99.9% (metals basis) | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
23985 | |
99.9% |
7440-57-5 | |
(metals basis) | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
CHEBI:30050 |
Safety and Handling
missing translation for 'einecsNumber' : 231-165-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only