missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Round-Base Test Tubes

Providing high resistance to centrifugation up to 3000g with a suitable adaptor in a balanced rotor. Gosselin™ Round-Base Test Tubes are available in either polystyrene or polypropylene.

Brand:  Gosselin™ TH55-03

 View more versions of this product

Product Code. 11783779

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For laboratory use
  • All tubes are CE + IVD marked
  • Available with or without closure
  • Compatible with automation systems
  • Available in sterile or non-sterile (sterility provided by electron beam treatment at 10kGy)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE, IVD, EC N°1935/2004, ISO 11137
Natural
55 mm
Winged plug
3000 Carton
12.80 mm
Round Bottom Tube
LDPE
3 mL
Polystyrene
750BG
Sterile
Wing Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.