missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Autoclavable Bags
For autoclaving contaminated products safely
Brand: Corning™ SA09-05
Code : NEW
Additional Details : Weight : 0.02000kg
Description
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent
Specifications
Autoclavable | |
Natural | |
Biohazard symbol | |
40 μm | |
30 cm | |
50 cm |
9 L | |
Clinical, dairy, environment, industries, microbiology | |
HDPE | |
11.81 in. | |
19.68 in. |