Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
Brand: A&D Weighing GP-20K-EC
This item is not returnable.
View return policy
Description
- EC Type approved
- Swing arm design or separate display and platform design
- IP65 protected
- Durable aluminum alloy & stainless steel construction
- Easy calibration with internal calibration mass
- SHS response speed of only 1.5 seconds
- GLP, GMP and ISO compliant
- RS-232C interface
- Comparator output and analog output (optional)
- Large Vacuum Fluorescent display
- Data memory function
- Counting function with ACAI (Automatic Counting Accuracy Improvement)
- Smart range function (GP-32K/102K/32KS)
- Comparator function
- Underhook weighing for thermal and magnetic samples as well as for density calculation
- Animal weighing function
Specifications
21 kg | |
0.1 g | |
Industrial Precision Balance |
VFD | |
1.5 seconds |