Learn More
GPR50 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19762-1-AP-20UL
 View more versions of this product
Product Code. 16859244
  • £135.00 / 20µL
Explore more special offers
Description
Description
Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
Specifications
Specifications
GPR50
Polyclonal
Unconjugated
GPR50
G protein coupled receptor 50, GPR50, H9, Mel1c, Melatonin related receptor
Rabbit
Antigen Affinity Chromatography
RUO
14765, 9248
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88495, Q13585
GPR50
Peptide
20 μL
Primary
Mouse, Human