missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Grab-It™ Swab Holder

Ideal for holding cloth swabs for cleaning hard-to-reach places.

Brand:  Sampling Systems™ 8420A-1300

 View more versions of this product

Product Code. 15859511

  • £59.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lightweight design
  • Manufactured from aluminum and plastic
  • Available in two lengths, 750 or 1300mm
Specifications

Specifications

Swab Holder
Lightweight
1300 mm
Aluminum, Plastic
Cleaning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers