missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Grab-It™ Swab Holder

Ideal for holding cloth swabs for cleaning hard-to-reach places.

Brand:  Sampling Systems™ 8420A-750

Additional Details : Weight : 0.99600kg

 View more versions of this product

Product Code. 15869511

  • £33.75 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Lightweight design
  • Manufactured from aluminum and plastic
  • Available in two lengths, 750 or 1300mm
Specifications

Specifications

Swab Holder
Lightweight
750 mm
Aluminum, Plastic
Cleaning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers