missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Grace's Insect Medium, supplemented
Grace's Insect Medium, supplemented
Brand: Gibco™ 11605045
Code : 50
Additional Details : Weight : 0.50000kg
Description
Grace's Insect Medium, Supplemented, was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Grace's Supplemented Insect Medium is now widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21.Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ media options for a range of insect cell culture applications.
Formulation
This Grace's Insect Medium contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate. The complete formulation is available. Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate not commonly found in the unsupplemented version.
Dual site cGMP manufacturing and quality system
Gibco™ Grace's Supplemented Insect Medium is manufactured at a cGMP-compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Grace's Supplemented Insect Medium made in our Grand Island facility (11605-094). This facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 and 9001 standards.
Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate, and is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. Grace's Supplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.
Specifications
2 to 8°C | |
Insect Cell | |
Grace's Insect Medium | |
500 mL |
Grace's Insect Medium, 500mL, Supplemented, Insect Cells, Sf21, Sf9 Cell Line, L-Glutamine, Calcium, Magnesium Inorganic Salt Type, Insect Hemolymph-Free, Lactalbumin Hydrolysate, Yeastolate Supplements, 2 to 8°C Storage Temperature | |
Liquid | |
Insect cell culture | |
For research use only. Not for use in diagnostic procedures |
For Research Use Only. Not for use in diagnostic procedures.