missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Grace's Insect Medium, supplemented
Grace's Insect Medium, supplemented
£57.00 - £441.00
Specifications
Content And Storage | 2 to 8°C |
---|---|
Cell Type | Insect Cell |
Form | Liquid |
Product Type | Grace's Insect Medium |
For Use With (Application) | Insect cell culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11590396
|
Gibco™
11605045 |
500 mL |
£57.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11500406
|
Gibco™
11605086 |
10 x 500 mL |
£441.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Grace's Insect Medium, Supplemented, was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Grace's Supplemented Insect Medium is now widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21.Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ media options for a range of insect cell culture applications.
Formulation
This Grace's Insect Medium contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate. The complete formulation is available. Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate not commonly found in the unsupplemented version.
Dual site cGMP manufacturing and quality system
Gibco™ Grace's Supplemented Insect Medium is manufactured at a cGMP-compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Grace's Supplemented Insect Medium made in our Grand Island facility (11605-094). This facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 and 9001 standards.
Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate, and is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. Grace's Supplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.
Specifications
2 to 8°C | |
Liquid | |
Insect cell culture |
Insect Cell | |
Grace's Insect Medium | |
For research use only. Not for use in diagnostic procedures |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title