missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Smooth Filter Paper Discs, Grade 3-HW
Use for routine qualitative analyses and technical applications. Sartorius™ Smooth Filter Paper Discs, Grade 3-HW can be used for pressure and vacuum filtration. Ultra-pure filters are available in filter disc format in a variety of diameters.
£7.70 - £76.40
Specifications
Format | Disc |
---|---|
Material | Cotton Linters, Refined Pulp |
Particle Retention | 8 to 12 μm |
Filtration Speed | 20μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
10357261
|
Sartorius™
FT-3-303-100 |
100 mm |
£15.65
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10498932
|
Sartorius™
FT-3-303-110 |
110 mm |
£14.50
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10397401
|
Sartorius™
FT-3-303-120 |
120 mm |
£14.50
Pack of 100 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10375171
|
Sartorius™
FT-3-303-125 |
125 mm |
£17.15
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10150611
|
Sartorius™
FT-3-303-150 |
150 mm |
£14.50
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10239244
|
Sartorius™
FT-3-303-015 |
15 mm |
£64.80
Pack of 1000 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
10079670
|
Sartorius™
FT-3-303-185 |
185 mm |
£22.25
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10643962
|
Sartorius™
FT-3-303-240 |
240 mm |
£32.00
Pack of 100 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
10141141
|
Sartorius™
FT-3-303-270 |
270 mm |
£38.50
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10039650
|
Sartorius™
FT-3-303-320 |
320 mm |
£52.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10652592
|
Sartorius™
FT-3-303-385 |
385 mm |
£76.40
Pack of 100 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
10045864
|
Sartorius™
FT-3-303-040 |
40 mm |
£11.30
Pack of 100 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
10295701
|
Sartorius™
FT-3-303-050 |
50 mm |
£10.95
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10767271
|
Sartorius™
FT-3-303-055 |
55 mm |
£7.70
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10431812
|
Sartorius™
FT-3-303-060 |
60 mm |
£10.95
Pack of 100 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10101191
|
Sartorius™
FT-3-303-070 |
70 mm |
£7.70
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10602592
|
Sartorius™
FT-3-303-080 |
80 mm |
£13.35
Pack of 100 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
10273331
|
Sartorius™
FT-3-303-085 |
85 mm |
£8.70
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10029640
|
Sartorius™
FT-3-303-090 |
90 mm |
£13.20
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10441812
|
Sartorius™
FT-3-303-095 |
95 mm |
£13.20
Pack of 100 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
10158771
|
Sartorius™
FT-3-303-045 |
45 mm |
£7.70
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
These smooth filter paper discs are used for routine qualitative analyses, such as clarification and determination of substances. The discs feature a center hole for technical applications. Filters are wet-strengthened, so they are suitable for pressure or vacuum filtration.- Made of refined pulp and cotton linters with a α-cellulose content greater than 95%
- Pure, technical filter paper with an ash content of 0.1 to 0.15%
- Grade 3-HW filters feature the following characteristics:
- Medium to wide pore for a moderately-fast filtering rate
- Wet burst resistance of 15kPa
- Basis weight of 65g/m2
- Non-sterile and bulk packed
Specifications
Disc | |
8 to 12 μm | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
0.14 mm |
Cotton Linters, Refined Pulp | |
20μm/sec. | |
3HW | |
Qualitative, Technical, Smooth |