missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 501 Chromatography Paper

Fisherbrand™ Grade 501 Chromatography Paper is standard grade chromatography paper for routine or simple analysis.

Brand:  Fisherbrand™ 0501F00026

97.13 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11718742

  • £111.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Grade 501
  • Standard chromatography paper for routine and simple analysis
Specifications

Specifications

0.19 mm
Sheet
Paper
145 mm/30 min.
Standard Chromatography Paper for Routine and Simple Analysis
501
200 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Grade 501 Chromatography Paper > Length: 200mm; Width: 200mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.