missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 907 Germination and Seed Test Papers, Plain Disc
For germination assays of seed specimens according to various methods such as Neeb and Jacobsen.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Wetstrength paper 160g/m²
- Absorbent
- Free from chlorine
- pH must be between 6.0 and 7.5
- For small seeds such as grasses and flowers
- White in color
Specifications
160 g/m2 | |
Paper | |
100 | |
0.38 mm | |
85 mm |
White | |
Germination assays of seed specimen | |
6 to 7.5 | |
Germination and Seed Test Paper |