missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 907 Germination and Seed Test Papers, Plain Disc

For germination assays of seed specimens according to various methods such as Neeb and Jacobsen.

£10.81 - £23.30

Specifications

Basis Weight 160 g/m2
Color White
Material Paper
For Use With (Application) Germination assays of seed specimen
No. per Pack 100
View More Specs

Products 2
Product Code Brand Diameter (Metric) No. per Pack Price Quantity & Availability  
Product Code Brand Diameter (Metric) No. per Pack Price Quantity & Availability  
11759905
View Documents
Fisherbrand™
0907A00001
85 mm 100 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11769905
View Documents
Fisherbrand™
0907A00002
90 mm 100 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Wetstrength paper 160g/m2
  • Absorbent
  • Free from chlorine
  • pH must be between 6.0 and 7.5
  • For small seeds such as grasses and flowers
  • White in color
Specifications

Specifications

160 g/m2
Paper
100
0.38 mm
White
Germination assays of seed specimen
6 to 7.5
Germination and Seed Test Paper
Videos
SDS
Documents

Documents

Product Certifications